missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Disposable Cuvettes
Choice of polystyrene or methacrylate
$247.00 - $382.00
Specifications
| Disposable | Single-use |
|---|---|
| Length (Metric) | 12mm |
| Quantity | 500 Case |
| Path Length | 10 mm |
| Height (Metric) | 45 mm |
| Catalog Number | Capacity (Metric) | Type | Material | Wavelength Range | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Capacity (Metric) | Type | Material | Wavelength Range | Price | Quantity & Availability | |||
|
14955129
|
4.5 mL | Cuvette | Polystyrene | 340 to 750 nm |
Case of 500 Each for $247.00
|
|
|||
|
14955126
|
4.5 mL | Standard Cuvette | PMMA | 285 to 750 nm |
Case of 500 Each for $382.00
|
|
|||
|
14955130
|
4.5 mL | Cuvette | PMMA | 285 to 750 nm |
Case of 500 Each for $381.00
|
|
|||
|
14955125
|
4.5 mL | Standard Cuvette | Polystyrene | 340 to 750 nm |
Case of 500 Each for $267.00
|
|
|||
|
14955127
|
1.5 mL | Semi-micro Cell | Polystyrene | 340 to 750 nm |
Case of 500 Each for $275.00
|
|
|||
|
14955128
|
1.5 mL | Semi-micro Cell | Methacrylate | 285 to 750 nm |
Case of 500 Each for $359.00
|
|
|||
Description
Fisherbrand™ Disposable Cuvettes are available in choice of polystyrene or methacrylate. Select polystyrene cuvets for assays in visible spectral range (340 to 750nm); for accuracy throughout UV/Vis range (285 to 750nm), choose cuvets made from a superior grade of methacrylate.
- Disposable 10mm pathlength cuvets designed for visible and UV/Vis assays
- Choice of 4.5 and 1.5mL sample capacity
- Clear four-sided cuvets are ideal for fluorimetry and nephelometry, as well as spectrophotometry
- Scale down assay volumes and save on reagents by using semimicro cuvets with capacity to 1.5mL
- Molded-in arrow indicates direction of transmission on standard and semimicro cuvets—helping assure uniformity
- Each case contains cuvets with same mold impression number—assuring precision between samples—standard deviation is <±0.005 extinction units between cuvets
Order Info
Packaged 100 cuvets per stackable tray; five trays to a case. Outside dimensions: 0.47L x 0.47W x 1.77 in. H (12 x 12 x 45mm).
Specifications
| Single-use | |
| 500 Case | |
| 45 mm | |
| 12 mm |
| 12mm | |
| 10 mm | |
| Spectrophotometers, Fluorometers, Nephelometers | |
| For assays in visible spectral range, accuracy throughout UV/Visible range |