missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Disposable Controlled-Drop Pasteur Pipets
Perfect for blood typing and cross-matching procedures in blood bank laboratories
$296.83 - $296.83
Specifications
| Diameter (Metric) Inner | Tip Opening: 1.20 to 1.65 mm |
|---|---|
| Diameter (Metric) Outer | 6.95 mm (top) |
| For Use With (Application) | For blood typing and cross-matching procedures in blood bank laboratories |
| Length (English) | 5.75 in. |
| Volume (Metric) | 2 mL |
Description
Perfect for blood typing and cross-matching procedures in blood bank laboratories
- Deliver 22 to 26 drops per mL
- Correspond to drop size from antisera reagent bottle droppers
- Can be cotton plugged (plugs not included)
Order Info
250 Pipets per pack, four packs per case.
Specifications
| Tip Opening: 1.20 to 1.65 mm | |
| For blood typing and cross-matching procedures in blood bank laboratories | |
| 2 mL | |
| Controlled Drop Pasteur Pipet |
| 6.95 mm (top) | |
| 5.75 in. | |
| 14.6 cm | |
| Single-use |