missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Disposable Auto-Analyzer Cups
Description
- Precision-molded clear polystyrene
- Graduated with round or conical inside bottoms to maximize sample uptake
Specifications
Specifications
| For Use With (Equipment) | Kodak Ektachem E400 and E700, Centrifichem and Technicon Analyzer |
| Disposable | Single-use |
| Color | Clear |
| Product Type | Auto Analyzer Cup |
| Material | Polystyrene |
| Volume (Metric) | 0.5 mL |
| Graduated | Yes |
| Shape | Round or Conical |