missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Disposable Auto-Analyzer Cups

Catalog No. 025446
Change view
Click to view available options
Volume (Metric):
2 mL
For Use With (Equipment):
Hitachi 700 Series, Nova, IL 1800 Analyzers
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) For Use With (Equipment)
025446 2 mL Hitachi 700 Series, Nova, IL 1800 Analyzers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 025446

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifically designed for use with most clinical instrumentation

  • Precision-molded clear polystyrene
  • Graduated with round or conical inside bottoms to maximize sample uptake

Specifications

For Use With (Equipment) Hitachi 700 Series, Nova, IL 1800 Analyzers
Disposable Single-use
Material Polystyrene
Color Clear
Product Type Auto Analyzer Cup
Volume (Metric) 2 mL
Shape Round or Conical
Graduated Yes