Learn More
Fisherbrand™ Disposable Animal Feeding Needles
For critical applications requiring single-use feeding needles.
$310.45 - $451.66
Specifications
Product Type Malleable Animal Feeding Needle
Disposable Single-use
For Use With (Application) For critical applications requiring sterile, single use feeding needles
Sterility Sterile
View More Specs
Products 3
Catalog Number Gauge Length (English) Length (Metric) Price Quantity & Availability  
Catalog Number Gauge Length (English) Length (Metric) Price Quantity & Availability  
0120888
View Documents Promotion Details
18G 2 in. 5.1 cm
Pack of 100 Each for $310.45
Only null left
0120889
View Documents Promotion Details
15G 3 in. 7.6 cm
Pack of 100 Each for $451.66
Only null left
0120887
View Documents Promotion Details
20G 1.5 in. 3.8 cm
Pack of 100 Each for $451.66
Only null left
Description
Description
  • Nontoxic, nonpyrogenic
  • Malleable stainless-steel tubing and silicone tip
  • Plastic Luer hubs
  • Sterile, packaged in Tyvek™ pouches
  • Five per pouch, 20 pouches per box
Specifications
Specifications
Malleable Animal Feeding Needle
Single-use
For critical applications requiring sterile, single use feeding needles
Sterile