missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Dispenser Tips for Handystep™ S Repeating Pipetter

Catalog No. 13668717
Change view
Click to view available options
Volume (Metric):
Tip Adapters for 25 and 50 mL
0.1 mL
0.5 mL
1 mL
1.25 mL
2.5 mL
5 mL
10 mL
12.5 mL
25 mL
50 mL
Sterility:
Non-sterile
Sterile
22 product options available for selection
Product selection table with 22 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility
13668717 12.5 mL Sterile
13668700 0.1 mL Non-sterile
13668701 0.5 mL Non-sterile
13668702 1 mL Non-sterile
13668703 1.25 mL Non-sterile
13668704 2.5 mL Non-sterile
13668705 5 mL Non-sterile
13668706 10 mL Non-sterile
13668707 12.5 mL Non-sterile
13668708 25 mL Non-sterile
13668709 50 mL Non-sterile
13668710 0.1 mL Sterile
13668711 0.5 mL Sterile
13668712 1 mL Sterile
13668713 1.25 mL Sterile
13668714 2.5 mL Sterile
13668715 5 mL Sterile
13668716 10 mL Sterile
13668718 25 mL Sterile
13668719 50 mL Sterile
13668720 Tip Adapters for 25 and 50 mL Non-sterile
13668721 Tip Adapters for 25 and 50 mL Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 22 options available.
22 options
Catalog No. 13668717 Supplier Fisherbrand™ Supplier No. 702997
Only null left

Manufactured from high-quality virgin plastics for use with most standard repeating pipettes in a wide variety of applications. The tips are made without chemical additives to prevent reagent contamination.

Manufactured from high-quality virgin plastics for use with all standard repeating pipettes in a wide variety of applications. The tips are made without chemical additives to prevent reagent contamination.

25 mL and 50 mL tips require the use of an adapter. One reusable adapter supplied in each package of 25 mL and 50 mL tips.

  • Precision seal between the piston and cylinder ensure smooth operation, increased accuracy and precision
  • Extended piston design minimizes dead volume and maximizes fluid utilization
  • Made entirely from high-quality virgin, high density polyethylene and polypropylene (piston of 0.1 mL tip is liquid crystal polymer-LCP)

Available sterile or nonsterileFits these models but not limited to:

  • BRAND™ HandyStep™, HandyStep™ S, HandyStep™ electronic, HandyStep™ touch, HandyStep™ touch S
  • All previous Fisherbrand Repeating Pipettes
  • Rainin AutoRep™ M, AutoRep™ S, AutoRep™ E
  • Eppendorf Multipette™ 4780
  • Gilson Repetman™
  • DLAB StepMate
  • Scilogex Repeater

Specifications

Volume (Metric) 12.5 mL
Volume (Metric) Dispensing 2500 to 12500μL
Filtered Non-filtered
Sterility Sterile
Color Translucent
Autoclavable Not Autoclavable
Precision ≤0.3 to 0.2%
Accuracy ±≤1.5 to 0.3%
Clean Claims Free from endotoxins, DNA, RNase, DNase, and ATP
Sterilization Method Beta
Format Individually Wrapped
Tip Style Positive Displacement
Quantity 100/Pk.
Product Type Syringe Tips
Disposable Single-use
For Use With (Equipment) Most repeating pipette models (see Description), Fisherbrand HandyStep S
Graduated Yes
Material HDPE, Polypropylene
Graduations Printed
Show More Show Less