missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Cytology Microslides

Catalog No. 12550019
Change view
Click to view available options
Thickness (Metric):
1 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thickness (Metric)
12550019 1 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12550019

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Use for cancer screening, hormonal reading and microbiological classification

  • Microslides with frosted ends
  • Clear corrosion-resistant glass with flat surfaces and smooth ground edges
  • Have letters V (vaginal), C (cervical), and E (endocervical) etched on reverse side
  • Hand-picked, precleaned, ready to use in one-at-a-time dispenser boxes of 72 slides

Order Info

Size: 75 x 25mm. Thickness, 1mm.

Specifications

Type Cytology Slide
Thickness (Metric) 1 mm
For Use With (Application) For cancer screening, hormonal reading and microbiological classification