Learn More
Fisherbrand™ Curved Tapered Flat-Tip Forcep
Curved tip for added visibility
Supplier:  Fisherbrand™ PL12
Catalog No. 16-100-111
Only null left
Explore available promotions
Description
Description
Use for handling labels or SMD components.
  • Made of antimagnetic stainless steel
  • Overall length is 4.5 in., L: 4.5 in. (115mm); W: 0.375 in. (9.5mm); H: 0.06 in. (1.5mm)
Specifications
Specifications
Curved, Tapered, Flat
Stainless Steel
Forceps
4.5 in.
115 mm
Dissection, research, laboratories, small-parts handling
Product Suggestions
Product Suggestions