Learn More
Fisherbrand™ Commode Specimen Collector
Catalog No. 22363149
Change view
Click to view available options
Quantity:
100/Cs.
Catalog No. Quantity
22363149 100/Cs.
1 options
Catalog No. 22363149
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
One-piece unit features easy-to-read imprinted graduations, instruction, and convenient pouring spout
  • Made of durable plastic
  • System components fit standard toilets
  • Easy-to-read graduations to determine specimen volume
Specifications
Product Type Commode Specimen Collection System
Includes Graduations on container, convenient pouring spout, instruction
Material Plastic
Quantity 100/Cs.