missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Clamp Swivel Holder

Catalog No. 02215465
Change view
Click to view available options
Size (Metric) Grip:
0 to 19 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (Metric) Grip
02215465 0 to 19 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02215465 Supplier Fisherbrand™ Supplier No. 80862771

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Two rod holders with center swivel capacity allow tilting of clamps at any angle in parallel planes.

Two rod holders with center swivel capacity allow tilting of clamps at any angle in parallel planes.

  • Nickel-plated zinc construction
  • Outside adjustment screw allows close proximity between items being held
  • Grips rods from 0 to 19mm (0 to 0.75 in.)

Specifications

Quantity 1 Ea.
Type Clamp Swivel Holder
Material Nickel-plated Zinc
Holds Rods, 0 to 0.75 in. (0 to 19 mm)
For Use With (Application) Holding rods from 0 to 0.75 in. (0 to 19mm)
Size (Metric) Grip 0 to 19 mm
Size (English) Grip 0 to 0.75 in.
Jaw Type Swivel