missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Cheesecloth Miniwipes, 200 pk.

Catalog No. 0666528
Change view
Click to view available options
Dimensions (L x W):
7 x 14 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
0666528 7 x 14 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 0666528
Edge

missing translation for 'validMainframeMsg'

Each wipe is a folded 4 in. (10.2cm) square pad that unfolds to approximately 7 x 14 in.

Fisherbrand™ Miniwipes can be used on microscopes, optical surfaces, glassware, pH electrodes. Each wipe is a folded 4 in. (10.2cm) square pad that unfolds to approximately 7 x 14 in.

  • Certified 100% reagent-grade cotton
  • Will not scratch sensitive surfaces
  • Absorbs 6 times weight
  • Size: 7 x 14 in. folded to 4 in. sq.

Order Info

Box dimensions: 4.5L x 4.5W x 8 in. H (11 x 11 x 20cm).

Specifications

Product Type Cheesecloth Miniwipe
Length (English) 7 in.
Length (Metric) 18 cm
Width (English) 14 in.
Width (Metric) 36 cm
Material Cotton
Color White
Dimensions (L x W) 7 x 14 in.
For Use With (Application) For use to wipe cuvettes and microscopes, clean cameras, and dry delicate Glassware, syringes, pipettes, and pH electrode
For Use With (Equipment) Microscopes, optical surfaces, Glassware, pH electrodes
Sterility Non-sterile
Show More Show Less