missing translation for 'onlineSavingsMsg'
Learn More

Cello-Seal™ Grease

Catalog No. C601100
Change view
Click to view available options
Quantity:
6 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C601100 6 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C601100 Supplier Thermo Fisher Scientific Supplier No. FLC601100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Won't add to pressure in vacuum system

  • Low vapor pressure lubricant with consistency maintains consistency of petrolatum at 100°C (212°F)
  • May be blended with Cello-Grease™ Lubricant for heavier consistency
  • 100g jar

Specifications

Product Type Grease
Format Liquid
For Use With (Application) Won't add to pressure in vacuum system
For Use With (Equipment) Cello-Grease™ Lubricant
Boiling Point 250°C
Melting Point 90°C to 130°C
Color Amber
Packaging Glass Jar
Quantity 6 Pk.
Volume (English) 3.52 oz.