missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Sterile Cell Strainers
Description
Rapidly isolate primary cells for single-cell suspensions using Fisherbrand™ Cell Strainers. Sterile and ready to use for straining stem and primary cell samples. Disposable strainers feature universal fit for compatibility with most 50mL conical tubes. Available in three color-coded mesh sizes.
Rapidly isolate primary cells
- Fast, simple alternative to gauze filtration
- Reliably dissociates cells from clumps or primary tissues
- Consistently obtain a uniform single-cell suspension
- Ideal for preparation of:
- Single-cell suspensions of stem and primary (blood, tissue, cancer) cells
- Specimens for primary cell cultures and immunogens
- Freezing stocks
- Filtering agglutinative proteins produced in inactivation serum
- Strong nylon mesh with evenly spaced pores for consistent results
- Sterilized by gamma irradiation and non-cytotoxic to maintain sample integrity
- Includes an extended lip for easier handling with forceps
- Color-coded polypropylene frame with tab enables easy handling and identification
- Individually packed, ready to use and disposable
Specifications
Specifications
| Mesh Material | Nylon |
| Disposable | Single-use |
| Color | Yellow |
| Material | Polypropylene |
| Product Type | Cell Strainer |
| Sterility | Sterile |
| For Use With (Equipment) | Conical Tubes, 50 mL |
| Mesh Size | 100 μm |