missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Sterile Cell Strainers

Catalog No. 22363549
Change view
Click to view available options
Mesh Size:
40 μm
100 μm
Color:
Blue
Yellow
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mesh Size Color
22363549 100 μm Yellow
22363547 40 μm Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22363549
Only null left

Sterile and ready to use for straining stem and primary cell samples. Disposable strainers feature universal fit for compatibility with most 50mL conical tubes. Available in three color-coded mesh sizes.

Rapidly isolate primary cells for single-cell suspensions using Fisherbrand™ Cell Strainers. Sterile and ready to use for straining stem and primary cell samples. Disposable strainers feature universal fit for compatibility with most 50mL conical tubes. Available in three color-coded mesh sizes.

Rapidly isolate primary cells
  • Fast, simple alternative to gauze filtration
  • Reliably dissociates cells from clumps or primary tissues
  • Consistently obtain a uniform single-cell suspension
  • Ideal for preparation of:
    • Single-cell suspensions of stem and primary (blood, tissue, cancer) cells
    • Specimens for primary cell cultures and immunogens
    • Freezing stocks
    • Filtering agglutinative proteins produced in inactivation serum
Reliable and easy to use
  • Strong nylon mesh with evenly spaced pores for consistent results
  • Sterilized by gamma irradiation and non-cytotoxic to maintain sample integrity
  • Includes an extended lip for easier handling with forceps
  • Color-coded polypropylene frame with tab enables easy handling and identification
  • Individually packed, ready to use and disposable

Specifications

Mesh Material Nylon
Disposable Single-use
Color Yellow
Material Polypropylene
Product Type Cell Strainer
Sterility Sterile
For Use With (Equipment) Conical Tubes, 50 mL
Mesh Size 100 μm