missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Caution: Contains Formaldehyde Labels

Bright red fluorescent labels display the universal biohazard symbol for easy identification.
$10.83
Specifications
| Color | White, Black, Red, Fluorescent |
|---|---|
| Format | Label |
| Material | Paper |
| Product Type | Biohazard Warning Tape |
| Display Text | Contains Formaldehylde |
| Catalog Number | Mfr. No. | Quantity | Size (English) | Size (Metric) | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Size (English) | Size (Metric) | Price | Quantity & Availability | |||
18999928
![]() |
Fisherbrand™
LBH 16 |
320/Pk. | 0.87 x 3 in. | 2.2 x 7.62 cm |
Each for $10.83
|
|
|||
Description
Strong adhesive ensures that labels remain in place.
Compliance
Meet OSHA requirements.
Specifications
| White, Black, Red, Fluorescent | |
| Paper | |
| Contains Formaldehylde |
| Label | |
| Biohazard Warning Tape | |
| Biohazard |