missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Burn-up Bin™ Biohazard Waste Boxes
Store flat, assemble in seconds.
Manufacturer: Fisherbrand™ 797025
Description
Store flat, assemble in seconds.
- Corrugated cardboard—single use
- 2mil polyliner safely holds biohazard waste
- Tight-fitting lid has flap opening to accept material; to dispose, simply close lid and transfer filled box to incinerator
- Graphics and biohazard symbols readily identify contents, encourage proper use
Specifications
Floor model | |
13.3 cm | |
69 cm | |
Floor | |
11 gal. | |
Quick identification for non-hazardous waste paper products | |
12 in. | |
Cardboard | |
Biohazard Waste Box |
5.25 in. | |
12 x 12 x 27 in. (30 x 30 x 69 cm) | |
30 cm | |
30 cm | |
White | |
27 in. | |
Polyethylene | |
2 mm | |
12 in. |