Learn More
Fisherbrand™ Borosilicate Glass Disposable Serological Pipets with Wide Tip, Standard Length
Sterile and plugged, multipack
$625.28 - $694.48
Specifications
Certifications/Compliance ASTM E714
Material Borosilicate Glass
Sterility Sterile
Graduated Yes
Tip Style Wide
View More Specs
Products 2
Catalog Number Mfr. No. Volume (Metric) Negative Graduations Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Negative Graduations Price Quantity  
13-678-35A
Accessories View Documents
N/A 5 mL 1 mL
Case of 400 Each for $625.28
Only null left
13-678-35B
Accessories View Documents
N/A 10 mL 2 mL
Case of 400 Each for $694.48
Only null left
Description
Description
Sterile and plugged, multipack
Chemically inert pipets have negative graduations as well as bidirectional scales.
  • Blue markings are easy to read
  • Packaging meets requirements of small-, medium-, or large-volume use
  • Individually wrapped pipets come in transparent plastic/paper wrappers for easy identification and tear-through, and are further overpacked in handy shelf container/dispensers; ideal for small volumes requiring sterility
  • Multipack with resealable flap for larger volume use permits easy access to several pipets at a time without contaminating unused pipets
  • Outer packaging is recyclable
Compliance
Calibrated To Deliver/Blow Out, Color-coded packaging on all pipets, except bulk packs, allows easy identification by size using ISO™ standard colors.
Specifications
Specifications
ASTM E714
Sterile
Wide
Serological Pipet
Borosilicate Glass
Yes
Yellow
Single-use
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'