missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ BloodBloc™ Biohazard Wipes, Liners and Rolls
Description
Wipes and Liners are non-sterile and not intended for use on patients.
- Soft, white absorbent wipe with bright-orange polyethylene backing is impermeable to blood, body fluids and strong solvents including xylene, xylene substitutes, acetones, ketones, alcohols and formalin
- Nonwoven cellulose captures and retains fluid; nonlinting for use with delicate instrumentation such as blood counters
- Orange flexible impermeable plastic backing prevents soak-through of pathogens and chemicals to gloves which help minimize cross-contamination when lab personal open doors or touch surfaces
- Can also be used as mats and liners; rolls can be cut to desired length
- One time use only
Specifications
Specifications
| Product Type | Biohazard Roll |
| Length (English) | 100 ft. |
| Length (Metric) | 30.48 m |
| Width (English) | 16 in. |
| Width (Metric) | 40.64 cm |
| Absorbancy | Standard |
| Color | Orange backing |
| Disposable | Single-use |
| Dimensions (L x W) | 100 ft. x 16 in. |
| For Use With (Application) | Prevent soak through of blood, body fluids, tissue and chemicals to gloves and surfaces when handling tubes, specimens, plates. |
| Show More |