missing translation for 'onlineSavingsMsg'
Learn More

General Purpose Transfer Pipette, 6 in., 4.9 mL, Non-sterile
GREENER_CHOICE

Supplier:  Thermo Fisher Scientific 336

Catalog No. 1371199




Description

Description

Single squeeze draws up to 2.4 mL

Put an end to matching rubber bulb with glass pipette. Since entire unit is disposable, these pipettes prevent cross-contamination to samples and reduce airborne contamination to personnel. Molded in one piece from see-through low-density polyethylene.

  • Capacity: approximately 5 mL
  • Delivers 24 to 26 drops/mL
  • Unbreakable all-in-one pipettes eliminate the hazards of broken glass and exposure to infectious materials
  • Inert to biological fluids and most acids
  • Work well anywhere there is a need for quick, safe transfer of fluids
  • One squeeze draws from 2.0 to 3.3 mL (depending on size used)
  • All deliver repeatable drop sizes
  • Can be sealed and refrigerated for sample storage (seal with Cat. No. 02-678 Hemato-Seal™ Tube Sealant)
Specifications

Specifications

1.9 mL
23
No
5 mL
General Purpose Pipette
LDPE
Research
500/Pk.
15.4 cm
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions