missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Blood Bank Crossmatch Holders

Catalog No. p-177825
Change view
Click to view available options
Positions:
24
38
90
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Positions
148105 90
1481051 24
1481053 38
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 148105 Supplier Fisherbrand™ Supplier No. 1595654
Only null left

For typing, screening, and other blood bank applications.

  • Clear acrylic shelves with white bases and sides
  • Holds donor sample tubes as well as tubes for donor's serum and cell suspension, ABO, Rh, antibody grouping, and screening
  • Available in four sizes

24-tube capacity:
Holds four 16mm O.D. sample tubes and twenty 10mm O.D. crossmatch tubes

29-tube capacity:
Holds one 16mm sample tube, one 13mm pilot tube serum, and twenty-seven 10mm phenotype, genotype, and cross-match tubes

38-tube capacity:
Holds thirty-eight tubes for five sets of tubes with printed identification on top panel

  • Set 1: One 16mm patient, one serum tube, and one cell suspension tube
  • Set 2: Four 10mm-14mm forwarding group drives
  • Set 3: Three reverse grouping tubes
  • Set 4: Sixteen major cross match tubes
  • Set 5: Twelve minor cross match tubes

90-tube capacity:
Holds six 16mm O.D. donor sample tubes, eighty-four 10 to 13mm tubes for donor's serum and cell suspension, ABO, Rh, antibody grouping, and screening

Specifications

Autoclavable Not Autoclavable
Color Clear, Transparent, White
Holds Tubes (10 to 16 mm)
Material Acrylic
Positions 90
For Use With (Equipment) Blood bank crossmatch tubes
Dimensions (L x W x H) 13.25 x 5.25 x 2.5 in. (33.6 x 13.3 x 6.35 cm)
Length (Metric) 33.6 cm
Length (English) 13.25 in.
Width (English) 5.25 in.
Height (English) 2.5 in.
Diameter (Metric) Opening 10 to 16 mm
Product Type Tube Rack
Height (Metric) 6.35 cm
Width (Metric) 13.3 cm
Show More Show Less