missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Biohazard Labels
GREENER_CHOICE

Catalog No. p-192458
Change view
Click to view available options
Width (English):
1 in.
2 in.
5 in.
Length (English):
1 in.
3 in.
5 in.
Length (Metric):
2.5 cm
7.62 cm
12.7 cm
Quantity:
1000/Pk.
250/Pk.
500/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English) Length (English) Length (Metric) Quantity
15926 1 in. 1 in. 2.5 cm 1000/Pk.
15928 5 in. 5 in. 12.7 cm 250/Pk.
15927 2 in. 3 in. 7.62 cm 500/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 15926
Only null left

Bright red fluorescent labels display the universal biohazard symbol for easy identification.

  • Hazard communication labels ensure safety and efficiency
  • Identify equipment, containers, rooms, materials, experimental animals or combinations of these items that may contain potentially infectious agents
  • Easy peel-off labels with pressure-sensitive adhesive

Use in accordance with applicable federal and state regulations.

Specifications

Adhesive Type Pressure-sensitive
Color Fluorescent Red, Black
Format Label
Material Paper
Product Type Biohazard Warning Tape
For Use With (Application) Biohazard
Quantity 1000/Pk.
Dimensions (L x W) 1 in. x 1 in.
Length (English) 1 in.
Length (Metric) 2.5 cm
Width (English) 1 in.
Show More Show Less