missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Baume Plain Form Hydrometers

Available in a variety of models measuring Baume of syrups and other liquids heavier than water in various ranges from 0° to 70°

Supplier:  Fisherbrand™ B618040220

 View more versions of this product

Catalog No. 13201530


Only null left

Description

Description

  • 300mm (12 in.) long
  • Lead-free ballast and glass
  • Individually serialized
  • Temperature standardization: 60°F (15°C)
  • Hand inspected by our U.S. quality team
  • Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST

Order Info

Should you require an individual Thermometer Calibration Report, factory calibrated thermometers or custom calibration services are available.

Specifications

Chemical Identifiers

Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST. Hand inspected by U.S. quality team in exclusive triple accredited/registered ISO/IEC 17025:2005 and A2LA Accredited Calibration Laboratory; internationally recognized and accepted by the signatory economies of the ILAC/MRA and located in ISO 14001:2004 Environmental Management System registered facility.
300 mm
Hydrometers should be stored in a clean, dry, safe place, and should not be subjected to extreme temperatures.
Used in the chemistry, pharmacology, food and beverage, starch industries and more.

Specifications

±0.1°
Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST. Hand inspected by U.S. quality team in exclusive triple accredited/registered ISO/IEC 17025:2005 and A2LA Accredited Calibration Laboratory; internationally recognized and accepted by the signatory economies of the ILAC/MRA and located in ISO 14001:2004 Environmental Management System registered facility.
Hydrometers should be stored in a clean, dry, safe place, and should not be subjected to extreme temperatures.
Used in the chemistry, pharmacology, food and beverage, starch industries and more.
0.1°
300 mm
19 to 31°
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions