missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Cleanroom Badge Clips

Catalog No. 18001509
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
18001509 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18001509 Supplier Fisherbrand™ Supplier No. 18001509

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Secure badges or ID cards to garments. Fisherbrand™ Cleanroom Badge Clips are made of three inch poly for badges or garments with no pockets. These swivel clips are metal-free.

  • Ideal for visually identifying different divisions, departments, titles, duties, or access categories
  • Non-metal clips perfect for metal-free environments
  • Double-bagged to “Class 10” specifications, for use in critical environments
  • Contain no metal
  • 50/pack

Specifications

Color White
For Use With (Application) Environments that require no metal such as working closely with electronic component or medical devices
Material Poly