missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Autoclavable Biohazard Waste Bags

Catalog No. 14828245
Change view
Click to view available options
Capacity (English):
13 to 16 gal.
20 to 30 gal.
7 to 10 gal.
8 to 12 gal.
44 gal.
Dimensions (L x W):
23 x 19 in.
30 x 24 in.
35 x 25 in.
38 x 31 in.
48 x 37 in.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Dimensions (L x W)
14828245 8 to 12 gal. 30 x 24 in.
14828244 7 to 10 gal. 23 x 19 in.
14828246 13 to 16 gal. 35 x 25 in.
14828247 20 to 30 gal. 38 x 31 in.
14828248 44 gal. 48 x 37 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 14828245 Supplier Fisherbrand™ Supplier No. 14828245
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Dual tested compliant.

Dual tested compliant

 Autoclaving is a very common way to sterilize infectious waste through the use of high temperature, high pressure steam over a specific time period. Sterilized waste is less expensive to dispose. Dual-tested autoclave bags are ASTM D1922 Elmendorf Tear tested to exceed 480g of tear strength (MD and TD) and ASTM D1709 tested to exceed 165g dart drop strength.

  • Integral steam processing indicator that turns brown when autoclaved
  • Bags comply with US DOT 49CFR Sections 173-197 and specific state regulations
  • Operating temperature tolerance up to 285degreeFdegreeAvailable in various sizes
  • Bag with black print
  • Gauge: 2mil
Note: Not compliant with Delaware, which requires 3 mil; check state regulations.

Specifications

Capacity (English) 8 to 12 gal.
Color Red, Black Print
Dimensions (L x W) 30 x 24 in.
Length (Metric) 76.2 cm
Width (Metric) 61 cm
Autoclavable Autoclavable