missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Autoclavable Bottletop Dispensers

Catalog No. 1361515
Change view
Click to view available options
Volume (Metric):
1 to 10 mL
10 to 100 mL
Increments:
0.2 mL
2.0 mL
Accuracy:
±0.6% (±0.060mL)
±0.6% (±3.000mL)
Precision:
±0.2% (±0.020mL)
±0.2% (±0.200mL)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Increments Accuracy Precision
1361515 10 to 100 mL 2.0 mL ±0.6% (±3.000mL) ±0.2% (±0.200mL)
1361512 1 to 10 mL 0.2 mL ±0.6% (±0.060mL) ±0.2% (±0.020mL)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 1361515

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Calibration certificate

Provides smooth, effortless dispensing.

Provides smooth, effortless dispensing

 

  • Wetted parts are PTFE, FEP and borosilicate glass (allows instrument to be used with a wide variety of chemicals)
  • 7 volume ranges to choose from
  • Bubble-free dispensing
  • Effortless plunger movement
  • Disassembles for easy cleaning and servicing
  • Autoclavable at 121°C, 15 psi for 10 to 15 minutes
  • Each bottle top dispenser includes one 28mm, 32mm, 38mm, 40mm, 45mm bottle neck adaptor

 

Component Description

  • Piston: PTFE
  • Cylinder: Glass
  • Volume Adjustment Knob: Screw Type
  • Valve Housing: PTFE
  • Valve Assembly: Glass Ball & Seal
  • Discharge Assembly: PTFE
  • Delivery Tube: FEP
  • Inlet Tube: FEP, 13 in. (with extension)
  • Calibration: Individually calibrated and certified (in-lab by user is also possible)
  • Accuracy & Reproducibility: In accordance with ISO 8655 standards
  • Compability: Excellent with most reagents

 

Certifications

ISO 8655

Warranty

One year free replacement warranty against any manufacturing defects. The period of one year starts from the invoice date of Fisher Scientific to its customer.

Specifications

Accuracy ±0.6% (±3.000mL)
Autoclavable Autoclavable
Material Glass
Volume (Metric) 10 to 100 mL
Includes 13-in. inlet tube (with extension)
Increments 2.0 mL
Precision ±0.2% (±0.200mL)