Learn More
Bio Plas™ Astral™ Inoculation Device System
Economical, disposable inoculation system designed for safety and dependable sterility
 View more versions of this product
Catalog No. 05-664-83
Only null left
Explore available promotions
Includes:
Autoclavable handle with every case
Description
Description
Economical, disposable inoculation system designed for safety and dependable sterility
Calibrated using Evans Blue Dye spectrophotometric procedure and gravimetric method
  • Free of DNA, RNase, DNase, pyrogens and PCR inhibitors
  • Astral loops, needles, and Clone-Piks are packaged static free into individual tubular chambers, which are then hermetically sealed with aluminum film
  • A dust cover is heat-shrunk onto the unit before it is irradiation sterilized
  • Use on an individual basis, or for quantitative procedures such as sampling, urine counts, or serial dilutions
To use:
  • Puncture aluminum film with the handle, pushing down until it attaches to the loop, needle, or Clone-Pik
  • Withdraw handle; inoculation device is ready for many applications
Order Info
Supplied 10 racks of 96 to a case
Specifications
Specifications
Single-use
Astral Handle
Handle present
Black
Astral™ inoculation device system
Product Suggestions
Product Suggestions