Learn More
Fisherbrand™ ASTM™ Specific Gravity Hydrometers for Liquids Lighter than Water
Accuracy of hydrometer meets or exceeds ASTM™ E 100 specifications.
 View more versions of this product
Catalog No. 116033X
Only null left
Description
Description
Accuracy of hydrometer meets or exceeds ASTM™ E 100 specifications.
  • Use Cylinder 08-530K
  • Temperature of standardization: 60°F
Order Info
A certificate of calibration is available; simply add the letter C to the end of the catalog number when ordering.
Compliance
Calibrated according to ASTM™ E 126 specifications
Specifications
Chemical Identifiers
60°F
Specifications
107H
ASTM E126
Soda Lime Glass
0.850 to 0.900 ASTM106H-62
60°F
0.001
260 mm
Liquids heavier than Water
260 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Accessories