Learn More
Fisherbrand™ Arsine Generator
Catalog No. 01405
Change view
Click to view available options
Standard Taper Joint:
24/40, 12
Catalog No. Standard Taper Joint
01405 24/40, 12
1 options
Catalog No. 01405
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Low-extractable borosilicate glass construction
  • Absorbance of complex is proportional to arsenic concentration
  • Easily measured with colorimeter or spectrophotometer
  • Samples are prepared in interchangeable flasks and are added, one after another, onto scrubber/absorber units
  • Ball-and-socket joint makes it easy to clean or recharge scrubber
  • Joints require no grease, eliminating a contamination source
Specifications
Product Type Arsine Generator
Standard Taper Joint 24/40, 12
Material Borosilicate Glass
Includes 125mL erlenmeyer flask, clamp, scrubber tube