Learn More
Fisherbrand™ Aluminum Dissecting Pan
Catalog No. 1381461
Change view
Click to view available options
Includes:
Wax
Without wax
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
1381461 Wax
1381460 Without wax
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 1381461 Supplier Fisherbrand™ Supplier No. 1381461
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Fisherbrand Aluminum Dissecting Pan is a major tool used in dissection across the globe.
  • Fisherbrand™ Aluminum Dissecting Pan is a world renown dissection tool designed for all a variety of dissection experiments
  • Major tool in dissection experiments
  • Made to last and feature safe, smooth, rolled edges for lasting rigidity and durability
  • Stack for compact storage
  • Rustproof
  • Available with or without black dissecting wax
  • Used in the fields of education and research
  • Made from ASTM™ standard aluminum
Order Info
Dimension: 11 x 7 x 1.5 in.
Specifications
Product Type Aluminum Dissecting Pan
Material Aluminum
For Use With (Application) General dissecting use
Includes Wax
Quantity 1 Ea.
Dimensions (L x W x H) 11 in. x 7 in. x 1.5 in.