Learn More
Fisherbrand™ Aluminum Baskets without Lids
Fisherbrand Aluminum Baskets without Lids are designed strong yet lightweight
 View more versions of this product
Catalog No. 1380721
Only null left
Description
Description
Fisherbrand Aluminum Baskets without Lids are designed strong yet lightweight
  • Fisherbrand™ Aluminum Baskets with Tilt-Top Lids are ideal for rinsing or storing tubes and confining small lab animals
  • Have many applications and various shapes and sizes are available
  • Diamond Flow-Through pattern allows for maximum penetration of sterilizer solution
  • Reinforced top, bottom and seams welded to frame plate
  • Rust and corrosion resistive
  • Baskets tilt at an optimum angle for easy dispening of test tubes and other labware and instruments
  • Tilt-cover doubles as angle supports
  • Also used to store animals
Specifications
Chemical Identifiers
No
Aluminum
Aluminum Basket
Not Autoclavable
Sterile
Specifications
No
Aluminum
Sterile
15.2 cm
6 in.
Aluminum Basket
15 cm
Not Autoclavable
Square
6 x 6 x 6 in.
6 in.
6 in.
15.2 cm
Product Suggestions
Product Suggestions