Learn More
Fisherbrand™ AFB✓™ Control Slides
Catalog No. 088015
Change view
Click to view available options
Quantity:
50/Pk.
Catalog No. Quantity
088015 50/Pk.
1 options
Catalog No. 088015
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AFB Check Slides are provided with a heat fixed attenuated positive Acid Fast organism and a negative Acid Fast organism with one additional circular area for your unknown specimen.
The AFB Check Slide has two circular shaped areas with heat-fixed attenuated smears of Gram-positive M. kansasii and Gram-negative E. coli, and a blank oval area for specimen examination.
Specifications
Quantity 50/Pk.