Learn More
Fisherbrand™ ACA Labels
GREENER_CHOICE
Determine equipment status at a glance.
 View more versions of this product
Catalog No. 15945
Only null left
Description
Description
  • Designed for use with Automated Clinical Analyzers (ACA)
  • Brightly colored, pressure-sensitive labels allow operators to check if equipment is calibrated or not
  • Color-coded labels: Red for “Not Calibrated—Do Not Use”; Green for “Ready for Use”; and Yellow for “Calibrated—Not in Use”
  • Paper labels may be written on
Specifications
Chemical Identifiers
Label
Paper
Automated clinical analyzers (ACA)
1000/Pk.
Specifications
Yellow
Paper
Calibrate-Not In Use
1000/Pk.
4.8 cm
Label
ACA Label
Automated clinical analyzers (ACA)
1.87 in.
1.87 in.
Product Suggestions
Product Suggestions