missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Absorption Ultra Micro Rectangular Cells with PTFE Lid

Catalog No. 14958138
Change view
Click to view available options
Capacity (Metric):
50 μL
100 μL
Description:
Center:15mm
Center:8.5mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Description
14958138 50 μL Center:8.5mm
14958136 100 μL Center:8.5mm
14958137 100 μL Center:15mm
14958139 50 μL Center:15mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14958138 Supplier Fisherbrand™ Supplier No. 14958138
Only null left

Includes

PTFE lid or stopper

Fit standard spectrophotometer cell holders

  • All cells are 12.5mm wide
  • 1, 5 or 10mm path length rectangular cells
  • Reduce reagent use with Ultramicro cells
  • Quartz windows for transmission from far ultraviolet through the infrared
  • Glass windows for routine work in ultraviolet region and visible region

Specifications

Type Ultra-micro Absorption Cell
Description Center:8.5mm
Capacity (Metric) 50 μL
Material Quartz
Closure Material PTFE
Width (Metric) Interior 2mm
Quantity Ea., 8 Ea./Cs.
Path Length 10 mm
Wavelength Range 200 to 2500 nm
Height (Metric) 45 mm
For Use With (Equipment) Standard Spectrophotometer Cell Holder
Width (Metric) 12.5 mm
For Use With (Application) Transmission from far ultraviolet through the infrared
Shape Rectangle
Show More Show Less