missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Absorption Ultra Micro Rectangular Cells with PTFE Lid
Description
Includes
PTFE lid or stopper
- All cells are 12.5mm wide
- 1, 5 or 10mm path length rectangular cells
- Reduce reagent use with Ultramicro cells
- Quartz windows for transmission from far ultraviolet through the infrared
- Glass windows for routine work in ultraviolet region and visible region
Specifications
Specifications
| Type | Ultra-micro Absorption Cell |
| Description | Center:15mm |
| Capacity (Metric) | 100 μL |
| Material | Quartz |
| Closure Material | PTFE |
| Width (Metric) Interior | 2mm |
| Quantity | Ea., 8 Ea./Cs. |
| Path Length | 10 mm |
| Wavelength Range | 200 to 2500 nm |
| Height (Metric) | 45 mm |
| Show More |