missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Semi-Micro Cuvette with PTFE lid

Catalog No. 14958108
Change view
Click to view available options
Capacity (Metric):
1500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
14958108 1500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14958108 Supplier Fisherbrand™ Supplier No. 14958108

Includes

PTFE lid

Contains quartz windows for transmission from far ultraviolet through the infrared.

  • Reduces the use of reagent with micro cells
  • Contains two clear windows made of quartz
  • With PTFE lid

Specifications

Type Semi-micro Cuvette
Capacity (Metric) 1500 μL
Material Quartz
Closure Material PTFE
Width (Metric) Interior 4mm
Quantity Ea.
Path Length 10 mm
Wavelength Range 200 to 2500 nm
Height (Metric) 49.5 mm
For Use With (Equipment) Standard spectrophotometer
Width (Metric) 12.5 mm
For Use With (Application) Transmission from far ultraviolet through the infrared
Shape Rectangle
Show More Show Less