missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Absorption Quartz Semi Micro Rectangular Cells with PTFE Lid, w/PTFE lid
Fit standard spectrophotometer cell holders
Specifications
| Type | Semi-micro Absorption Cell |
|---|---|
| Capacity (Metric) | 1000 μL |
| Material | Quartz |
| Closure Material | PTFE |
| Width (Metric) Interior | 4mm |
Includes: PTFE lid
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
14-958-125
|
Fisherbrand™
14958125 |
2/Pk., 8 Pk./Cs. |
N/A
|
|
|||||
Description
Fit standard spectrophotometer cell holders
- All cells are 12.5mm wide
- 10mm path length rectangular cells
- Reduce reagent use with Semimicro cells
- Quartz windows for transmission from far ultraviolet through the infrared
Specifications
| Semi-micro Absorption Cell | |
| Quartz | |
| 4mm | |
| 200 to 2500 nm | |
| Standard Spectrophotometer Cell Holder | |
| Transmission from far ultraviolet through the infrared |
| 1000 μL | |
| PTFE | |
| 10 mm | |
| 45 mm | |
| 12.5 mm | |
| Rectangle |