missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Absorption Quartz Micro Rectangular Cells with PTFE Lid
Fit standard spectrophotometer cell holders
$1,102.17
Specifications
| Type | Micro Absorption Cell |
|---|---|
| Material | Quartz |
| Closure Material | PTFE |
| Width (Metric) Interior | 2mm |
| Includes | 2 clear windows |
Includes: PTFE lid
| Catalog Number | Mfr. No. | Capacity (Metric) | Height (Metric) | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Height (Metric) | Price | Quantity & Availability | ||||
|
14958133
|
Fisherbrand™
14958133 |
300 μL | 25 mm |
Pack of 2 Each for $1,102.17
Case of 8 Pack for $8,817.36
|
|
||||
Description
Fit standard spectrophotometer cell holders
- All cells are 12.5mm wide
- 10mm path length rectangular cells
- Reduce reagent use with Micro cells
- Quartz windows for transmission from far ultraviolet through the infrared
Specifications
| Micro Absorption Cell | |
| PTFE | |
| 2 clear windows | |
| 10 mm | |
| Standard Spectrophotometer Cell Holder | |
| Transmission from far ultraviolet through the infrared |
| Quartz | |
| 2mm | |
| 2/Pk., 8 Pk./Cs. | |
| 200 to 2500 nm | |
| 12.5 mm | |
| Rectangle |