missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 96-Well Polypropylene Microplates
GREENER_CHOICE

Catalog No. p-156247
Change view
Click to view available options
Color:
Blue
Natural
Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
12565502 Natural
12565504 Yellow
12565505 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 12565502
Only null left

Ideal for storage—feature generous 500μL well volumes

  • Polypropylene
  • Minimum binding
  • Resistant to many reagents
  • Withstand temperatures from -80° to +121°C (-112° to +250°F)
  • Automation-friendly design optimized for HTS

Specifications

Format Standard
Array 8 x 12
Color Natural
Material Polypropylene
No. of Wells 96
Sterility Non-sterile
Well Shape Round
For Use With (Application) For storage feature generous 500μL well volumes