missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 96-Well Nonskirted PCR Plates

Catalog No. 14230233
Change view
Click to view available options
Color:
Blue
Green
Natural
Red
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14230233 Red
14230234 Blue
14230236 Yellow
14230235 Green
14230232 Natural
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 14230233 Supplier Fisherbrand™ Supplier No. 14230233

Compatible with most 0.2mL thermal cyclers and ABI PRISM™ 310, 3100, and 3700 capillary sequencers

  • Virgin polypropylene
  • Clear well bottom for sample visibility
  • Highly rigid plate deck
  • Raised rim design
  • Sealable using adhesive films and foils, heat seals, and cap strips
  • 300μL maximum volume when used with adhesive and heat seals
  • Cut-away corner and alphanumeric grid
  • Cleanroom produced

Specifications

Format Standard
Array 8 x 12
Color Red
Product Type PCR Microplate
Skirt Style Non-skirt
Material Polypropylene
Volume (Metric) Well 300 μL
No. of Wells 96
For Use With (Application) PCR, RT-PCR applications
Rimmed Yes
Well Color Clear
Show More Show Less