missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 384-Well Polystyrene Plates

Catalog No. p-153498
Change view
Click to view available options
Color:
Clear
Well Shape:
Rounded Square
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Well Shape
12565506 Clear Rounded Square
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12565506 Supplier Fisherbrand™ Supplier No. 235700

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Process four times as many samples as standard 96-well plates

Optimized for robotic instruments, plate readers and liquid handling systems.

  • Highly specified polystyrene
  • Ideal for low-volume assays such as colorimetric, enzymatic, cloning, replication, and array, as well as storage
  • Flat well bottom surface provides greater area for binding
  • Working volume: 10–100μL
  • 384-well microplates conserve samples and reagents, providing cost savings and greater screening productivity
  • Untreated
  • Non-sterile

Order Info

Accommodate 12-565-511 lid (sold separately).

Specifications

Format Standard
Lid Without Lid
Array 16 x 24
Color Clear
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 10 to 100 μL
Sterility Non-sterile
Surface Treatment Untreated
Well Shape Rounded Square
For Use With (Application) For low volume assays such as colorimetric, enzymatic, cloning, replication and array, as well as storage
Show More Show Less