missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ 384-Well DeepWell Polypropylene Microplates

Description
- Excellent for storing DMSO and other strong chemicals and solvents
- Well shape maximizes sample retrieval
- Compatible with robotics and automation
- Rounded square wells minimize wicking (capillary action)
- Barcoding available by arrangement
- Sealable with chemical-resistant well cap mats
- Working volume range: 5 to 240μL/well
Specifications
Specifications
| Array | 16 x 24 |
| Color | Natural |
| Product Type | 384-Well Microplate |
| Material | Polypropylene |
| No. of Wells | 384 |
| Sterility | Non-sterile |
| Well Shape | Square |
| For Use With (Application) | For storage of compounds and nucleic acid libraries |