missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Colored Labeling Tape, 1 in. wide

Useful for many labeling applications—will adhere to most surfaces and can be written on.
$11.79 - $27.74
Specifications
| Format | Tape |
|---|---|
| Material | Paper |
| For Use With (Application) | Waterproof, Oil and Acid resistant |
| Quantity | 3/Pk. |
| Temperature Range (English) | -10° to 250°F |
| Catalog Number | Mfr. No. | Color | Length (English) | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Length (English) | Price | Quantity & Availability | ||||
1590110J
![]() |
N/A | Copper | 14 yd. |
Pack of 3 Each for $25.83
Case of 4 Pack for $103.32
|
|
||||
1590110N
![]() |
N/A | Rose | 14 yd. |
Pack of 3 Each for $25.83
Case of 4 Pack for $103.32
|
|
||||
1590110Q
![]() |
N/A | Violet | 14 yd. |
Pack of 3 Each for $27.74
Case of 4 Pack for $110.96
|
|
||||
1590110B
![]() |
N/A | Yellow | 14 yd. |
Pack of 3 Each for $25.83
Case of 4 Pack for $103.32
|
|
||||
1590110A
![]() |
N/A | White | 14 yd. |
Pack of 3 Each for $25.83
Case of 4 Pack for $103.32
|
|
||||
1590110C
![]() |
N/A | Green | 14 yd. |
Pack of 3 Each for $25.83
Case of 4 Pack for $103.32
|
|
||||
1590110D
![]() |
N/A | Pink | 14 yd. |
Pack of 3 Each for $25.83
Case of 4 Pack for $103.32
|
|
||||
1590110E
![]() |
N/A | Red | 14 yd. |
Pack of 3 Each for $27.74
Case of 4 Pack for $110.96
|
|
||||
1590110F
![]() |
N/A | Orange | 14 yd. |
Pack of 3 Each for $27.74
Case of 4 Pack for $110.96
|
|
||||
1590110K
![]() |
Fisherbrand™
1590110K |
Gray | 14 yd. |
Pack of 3 Each for $11.79
Case of 4 Pack for $47.16
|
|
||||
1590110L
![]() |
N/A | Lavender | 14 yd. |
Pack of 3 Each for $11.79
Case of 4 Pack for $103.32
|
|
||||
1590110H
![]() |
N/A | Aqua | 14 yd. |
Pack of 3 Each for $11.79
Case of 4 Pack for $103.32
|
|
||||
1590110G
![]() |
N/A | Blue | 14 yd. |
Pack of 3 Each for $11.79
Case of 4 Pack for $103.32
|
|
||||
1590110M
![]() |
N/A | Lime Green | 14 yd. |
Pack of 3 Each for $11.79
Case of 4 Pack for $103.32
|
|
||||
1590125D
![]() |
N/A | Pink | 60 yd. |
Pack of 3 Each for $11.79
|
|
||||
1590125H
![]() |
N/A | Aqua | 60 yd. |
Pack of 3 Each for $11.79
|
|
||||
1590125L
![]() |
N/A | Lavender | 60 yd. |
Pack of 3 Each for $11.79
|
|
||||
1590125J
![]() |
N/A | Copper | 60 yd. |
Pack of 3 Each for $11.79
|
|
||||
1590125K
![]() |
N/A | Gray | 60 yd. |
Pack of 3 Each for $11.79
|
|
||||
1590125M
![]() |
N/A | Lime Green | 60 yd. |
Pack of 3 Each for $11.79
|
|
||||
1590125N
![]() |
N/A | Rose | 60 yd. |
Pack of 3 Each for $11.79
|
|
||||
1590125Q
![]() |
N/A | Violet | 60 yd. |
Pack of 3 Each for $11.79
|
|
||||
1590125A
![]() |
N/A | White | 60 yd. |
Case of 3 Each for $11.79
|
|
||||
1590125G
![]() |
N/A | Blue | 60 yd. |
Pack of 3 Each for $11.79
|
|
||||
1590125B
![]() |
N/A | Yellow | 60 yd. |
Pack of 3 Each for $11.79
|
|
||||
1590125C
![]() |
N/A | Green | 60 yd. |
Pack of 3 Each for $11.79
|
|
||||
1590125E
![]() |
N/A | Red | 60 yd. |
Case of 3 Each for $11.79
|
|
||||
1590125F
![]() |
N/A | Orange | 60 yd. |
Pack of 3 Each for $11.79
|
|
||||
Description
Colored Label Tape is useful for many labeling applications. It can be written on and will adhere to most surfaces.
- Strong, durable, and oil-, water-, and acid-resistant
- Excellent for color coding
- Withstand temperatures from -23° to +121°C (-10° to +250°F)
- Can be written on with pen, pencil, or permanent marker
- Stick to most surfaces including glass, plastic, and metal
- Remove cleanly and easily
- Width is 1 inch
Tapes are supplied without dispenser
Specifications
| Tape | |
| Waterproof, Oil and Acid resistant | |
| -10° to 250°F | |
| 1 in. |
| Paper | |
| 3/Pk. | |
| -23° to +121°C |