missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Semidry Blotting Apparatus

Catalog No. FBSDB2020
Change view
Click to view available options
Type:
Semi Dry Electroblotting System
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
FBSDB2020 Semi Dry Electroblotting System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FBSDB2020 Supplier Fisherbrand™ Supplier No. HEP1 XM FIS

Includes

Lid with anode, base with cathode, connecting knobs, power cords

Sleek, low-profile system for fast, efficient Western transfers

Transfers minigels in 15 to 30 minutes and full-sized gels in 30 to 60 minutes, with efficiencies of nearly 90%.

  • Multiple gels can be transferred by stacking gel/membrane/filter paper “sandwiches” Because plate electrodes are in direct contact with “sandwich,” a homogeneous electric field is created, ensuring even transfers System requires only minimal amounts of buffer (about 250mL) Low voltage—requires less than 1mA/cm2 of current; no external cooling is necessary
  • Features 20 × 20cm platinum-coated titanium anode and stainless-steel cathode

  • Transfers proteins from polyacrylamide electrophoresis gels onto hybridization membranes (Western transfers)
  • Ideal for membrane-based, solid-phase protein assays such as HIV testing, where viral antigens are assayed with patient antibodies
  • Use to detect trace proteins in complex mixtures and study protein interaction with other molecules (i.e., DNA)

Specifications

For Use With (Equipment) 25 x 25 x 5 cm Polyacrylamide Electrophoresis Gel
Type Semi Dry Electroblotting System
Includes Lid with 20 x 20cm Platinum coated Titanium anode, base with Stainless Steel cathode, connecting knob, power cords
Volume (English) Buffer 8.45 oz.
Volume (Metric) Buffer 250 mL
Dimensions (L x W x H) 9.75 x 9.75 x 2 in. (24.76 x 24.76 x 5.08 cm)
Length (English) 9.75 in.
Length (Metric) 24.76 cm
Width (English) 9.75 in.
Width (Metric) 24.76 cm
Height (English) 2 in.
Height (Metric) 5.08 cm
Amperage 1 mA/cm2
Show More Show Less