missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ DNA Sequencing Systems

Catalog No. FBSEQ3545
Change view
Click to view available options
Width (English) Gel:
12.99 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English) Gel
FBSEQ3545 12.99 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FBSEQ3545 Supplier Fisherbrand™ Supplier No. S3SXMFIS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Main chamber with aluminum backplate, clamps, removable lower buffer chamber, two notched glass plates, two blank glass plates, two sharktooth combs, one well comb, two spacer sets

Requires

High-voltage power supply

Run samples in all gel lanes without loss of data or questionable interpretation of the banding pattern

  • Integral anodized aluminum backplate distributes heat evenly across the entire gel surface to minimize band “smiling”
  • Screw clamps are mounted directly on the unit to ensure leakproof seal between gel sandwich and upper buffer chamber
  • Removable waste bottle allows for easy buffer disposal
  • Interlocking lids on upper and lower chambers provide safety

Specifications

For Use With (Application) Run samples in all gel lanes without loss of data or questionable interpretation of the banding pattern
Height (English) Exterior 19.68 in.
Height (Metric) Exterior 50 cm
Includes Main chamber with aluminum backplate, clamps, removable lower buffer chamber, 2 notched glass plates, 2 blank glass plates, 2 sharktooth combs, 1 well comb, 2 spacer sets
Length (English) Exterior 9.05 in.
Length (English) Gel 16.53 in.
Length (Metric) Exterior 23 cm
Length (Metric) Gel 41.98 cm
Product Type Dna Sequencing System
Volume (English) Buffer 27.05 oz.
Volume (Metric) Buffer 800mL
Width (English) Exterior 17.32 in.
Width (English) Gel 12.99 in.
Width (Metric) Gel 329.9 mm
Show More Show Less