Learn More
Fisherbrand™ DNA Sequencing Systems Sequencing Gel Caster
For FisherBiotech DNA Sequencing Systems
Supplier: Fisherbrand™ SGC 2
Description
- Cast any standard-size sequencing gel in less than three minutes
- Capillary action distributes gel smoothly, with no air bubbles
- Rugged acrylic casters come with two knobs for platform adjustment and 12 binder clamps
- 12W × 4.5 in.H (30 × 11.4cm)
- Platforms adjust for plates from 20 to 42cm wide and 48cm long
Specifications
| Dna sequencing systems | |
| 12.2 cm | |
| 7.8 to 16.5 in. | |
| 20 to 42 cm | |
| 18.8 in. | |
| 21 cm |
| 4.8 in. | |
| 12.2 cm | |
| 102 cm | |
| Sequencing Gel Caster | |
| 21 cm |