Learn More
Fisherbrand™ Vortex Mixer Head Cover
Catalog No. 02215383
Change view
Click to view available options
For Use With (Equipment):
Vortex Mixer
Catalog No. For Use With (Equipment)
02215383 Vortex Mixer
1 options
Catalog No. 02215383 Supplier Fisherbrand™ Supplier No. 80863000
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For use with Fisherbrand vortex mixers.
Also compatible with other manufacturers' models; contact your Fisher Scientific Customer Service Representative for details.
  • Rubber head (76mm) 02-215-382 is used with 76mm head cover 02-215-383 for mixing beakers, flasks, and irregular shaped objects
Specifications
Product Type Head Cover
For Use With (Equipment) Vortex Mixer