Learn More
Fisherbrand™ Vortex Mixer Head
Catalog No. 02215382
Change view
Click to view available options
For Use With (Equipment):
Vortex mixers
Catalog No. For Use With (Equipment)
02215382 Vortex mixers
1 options
Catalog No. 02215382 Supplier Fisherbrand™ Supplier No. 80862999
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For use with Fisherbrand vortex mixers.
Also compatible with other manufacturers' models; contact your Fisher Scientific Customer Service Representative for details.
  • Rubber head (76mm) 02-215-382 is used with 76mm head cover 02-215-383 for mixing beakers, flasks, and irregular shaped objects
Also compatible with other manufacturers' models; contact your Fisher Scientific Customer Service Representative for details.
  • Rubber head (76mm) 02-215-382 is used with 76mm head cover 02-215-383 for mixing beakers, flasks, and irregular shaped objects
Specifications
Product Type Head
Diameter (Metric) 76 mm
For Use With (Equipment) Vortex mixers