Learn More
Fisherbrand™ Vortex Mixer Accessories: Head and Head Cover
Catalog No. 02215381
Change view
Click to view available options
For Use With (Equipment):
Fisherbrand Vortex mixer
Catalog No. For Use With (Equipment)
02215381 Fisherbrand Vortex mixer
1 options
Catalog No. 02215381 Supplier Fisherbrand™ Supplier No. 80863007
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For use with Fisherbrand vortex mixers.
Also compatible with other manufacturers' models; contact your Fisher Scientific Customer Service Representative for details.
  • 3 in. Head permits easy-touch mixing of beakers and flasks
  • Head Cover slides over top of head—ideal for odd-shaped containers
  • Head With Rubber Cover is designed for mixing irregular shaped objects
Specifications
Product Type Head with Cover
Diameter (Metric) 76 mm
For Use With (Equipment) Fisherbrand Vortex mixer
Includes Head, Rubber cover