missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Traceable™ Visual Alarm Timer

Catalog No. 1464985
Change view
Click to view available options
Time Range:
Timing Capacity: 9999 min.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Time Range
1464985 Timing Capacity: 9999 min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1464985

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

AAA batteries (2), Integrated magnet, Stand, Clip

Ideal for repetitive tests

Ideal for repetitive tests. Fisher Scientific™ Traceable™ Visual Alarm Timer has a switch which allows user to select either an audible alarm or bright flashing LED visual alarm.

  • Counts down from 9999 minutes in one-minute increments
  • Countdown resolution is 1 second for times under 99 minutes
  • Automatic memory resets the display to the last programmed time
  • Thru-zero feature allows counting down, alarming, then counting up
  • Digits are 1cm (3/8 in.) high and readable from 3M (10 ft.)

Specifications

Product Type Benchtop Timer
Time Range Timing Capacity: 9999 min.
Alarms Audible/Visible
Accuracy 0.0001
Resolution 1 second to 1 minute
Display LED
Battery Type AAA
Weight (English) 0.18 lb.
Weight (Metric) 0.08 kg
Includes Two AAA batteries, integrated magnet, stand, clip
For Use With (Application) For repetative tests
Certifications/Compliance NIST
Show More Show Less