missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Traceable™ Refrigerator/Freezer Plus Thermometer

Catalog No. 15077721
Change view
Click to view available options
Temperature Range (Metric):
-50°C to +70°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Temperature Range (Metric)
15077721 -50°C to +70°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15077721
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Accurately monitors temperatures in freezers, water baths, heating blocks, incubators, and refrigerators with enclosed temperature-buffered sensor

Fisher Scientific™ Traceable™ Refrigerator/Freezer Plus Thermometer accurately monitors temperatures in freezers, water baths, heating blocks, incubators, and refrigerators with enclosed temperature-buffered sensor.

With 5mL round bottle. Vaccine bottle mimics vaccine temperature and fits in vaccine tray.
  • Probe is sealed in glycol solution
  • Bottle insulates sensor from rapid temperature changes when refrigerator door is opened
  • Monitors high/low readings overnight, on weekend or for any time period - a significant advantage over reading-only glass thermometers
  • Temperature range is -58 to 158°F and -50 to +70°C with a resolution of 0.1 degree and an accuracy of +/- 0.5C
  • Minimum/maximum memory
  • High and low settable alarms

Specifications

Certifications/Compliance ISO 17025, A2LA, NIST
Diameter (English) Probe 0.18 in.
Dimensions (L x W x H) 3.25 x 2.5 x 0.6 in. (8.3 x 6.35 x 1.5 cm)
For Use With (Application) Monitoring temperature in freezers, water baths, heating blocks, incubators and refrigerators
Height (English) 0.6 in.
Height (Metric) 1.5 cm
Includes Velcro™, magnetic strips, traceable certificate and AAA battery
Length (English) Cable 120 in.
Length (English) Probe 11.2 ft.
Length (Metric) 83 mm
Length (Metric) Cable 3.4 m
Length (Metric) Probe 3.17 cm
Material ABS Plastic
Resolution 0.1°C
Weight (English) 4 oz.
Weight (Metric) 110 g
Width (English) 2.5 in.
Product Type Digital Thermometer
Length (English) 3.25 in.
Max. Temperature (English) 158°F
Max. Temperature (Metric) 70°C
Min. Temperature (English) -58°F
Min. Temperature (Metric) -50°C
Temperature Range (English) -58°F to +158°F
Temperature Range (Metric) -50°C to +70°C
Calibrated Yes
For Use With (Equipment) Monitoring Temperature in Freezer
Accuracy ±0.5°C (±0.9°F) (between 0-50°C)
Display Digital
Width (Metric) 6.35 cm
Show More Show Less